N-(Ethoxycarbonyl)phthalimide ≥97%

Supplier: Thermo Scientific

Synonyms: Ethyl 1,3-dioxoisoindoline 2-carboxylate, N-Carbethoxyphthalimide

A14822.14 A14822.22
A14822.14EA 61 GBP
A14822.14 A14822.22
N-(Ethoxycarbonyl)phthalimide ≥97%
N-(Ethoxycarbonyl)phthalimide

Purity: 97% CAS No.: 22509-74-6 Molecular Formula: C11H9NO4 Formula Weight: 219.20 Melting Point: 78-84° Beilstein Reference: 196340

Formula: C₁₁H₉NO₄
MW: 219,20 g/mol
Melting Pt: 78…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00005893
CAS Number: 22509-74-6
EINECS: 245-048-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR