2-Chloro-3-methylquinoline ≥97%
Supplier: Thermo Scientific
|
Warning
|
H50492.03
H50492.06
H50492.03EA
32.1
GBP
H50492.03
H50492.06
2-Chloro-3-methylquinoline ≥97%
2-Chloro-3-methylquinoline
Purity: 97%
CAS No.: 57876-69-4
Molecular Formula: C10H8ClN
Formula Weight: 177.63
Melting Point: 82-84°
MDL No.: MFCD00100265
Formula:
C₁₀H₈ClN MW: 177,63 g/mol Melting Pt: 82…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00100265 CAS Number: 57876-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...