2-Pyridylacetonitrile ≥99%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
2-Pyridineacetonitrile
A13087.06
A13087.14
A13087.22
A13087.06EA
40.3
GBP
A13087.06
A13087.14
A13087.22
2-Pyridylacetonitrile ≥99%
2-Pyridylacetonitrile
Purity: 99%
CAS No.: 2739-97-1
Molecular Formula: C7H6N2
Formula Weight: 118.14
Flash Point: 93°(199°F)
Refractive Index: 1.5250
Melting Point: 23-25°
Boiling Point: 66-68°/1mm
Beilstein Reference: 111215
UN No.: 3439
MDL No.: MFCD00006346
Formula:
C₇H₆N₂ Boiling Pt: 66…68 °C (1 mmHg) Melting Pt: 23…25 °C Density: 1,059 Flash Pt: 93 °C (199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006346 CAS Number: 2739-97-1 EINECS: 220-364-6 UN: 3276 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...