3-Iodooxetane 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Iodo-1,3-epoxypropane, 1,3-Epoxy-2-iodopropane
H57302.MD
H57302.ME
H57302.MDEA
186
GBP
H57302.MD
H57302.ME
3-Iodooxetane 95%
3-Iodooxetane
Formula:
C₃H₅IO MW: 183,98 g/mol Boiling Pt: 166 °C Density: 2,14 Flash Pt: 54 °C (129 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD08544403 CAS Number: 26272-85-5 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...