3'-Methylbiphenyl-4-carboxylic acid 96%

Supplier: Thermo Fisher Scientific
Warning

H57426.03 H57426.06
H57426.03EA 31.7 GBP
H57426.03 H57426.06
3'-Methylbiphenyl-4-carboxylic acid 96%
3'-Methylbiphenyl-4-carboxylic acid
Formula: C₁₄H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03424590
CAS Number: 5728-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR