3-Amino-5-bromopyridine 98%

Supplier: Thermo Scientific
Warning

Synonyms: 5-Bromopyridin-3-amine, 5-Bromo-3-pyridylamine

H56566.03 H56566.06 H56566.14
H56566.03EA 21 GBP
H56566.03 H56566.06 H56566.14
3-Amino-5-bromopyridine 98%
3-Amino-5-bromopyridine
Formula: C₅H₅BrN₂
MW: 173,01 g/mol
Melting Pt: 65…69 °C
Storage Temperature: Ambient
MDL Number: MFCD01646060
CAS Number: 13535-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR