4-Fluoroindole ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Fluoro-1H-indole

H56011.MD H56011.03 H56011.06
H56011.MDEA 66.8 GBP
H56011.MD H56011.03 H56011.06
4-Fluoroindole ≥97%
4-Fluoroindole

Purity: 97% CAS No.: 387-43-9 Molecular Formula: C8H6FN Formula Weight: 135.14 Melting Point: 30-32° Boiling Point: 90°/0.4mm MDL No.: MFCD00055992

Formula: C₈H₆FN
MW: 135,14 g/mol
Boiling Pt: 90 °C (0,4 mmHg)
Melting Pt: 30…32 °C
Storage Temperature: Ambient
MDL Number: MFCD00055992
CAS Number: 387-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR