Ready to use microbiology media, plated, 90 mm
Supplier: E AND O LABORATORIES
PP0209-P090 PP0280-P090 PP0320-P090 PP1282-P090 PP1420-P090 PP1520-P090 PP1720-P090 PP2182-P090
EOLAPP0209-P090EA 12.7 GBP
EOLAPP0209-P090 EOLAPP0280-P090 EOLAPP0320-P090 EOLAPP1282-P090 EOLAPP1420-P090 EOLAPP1520-P090 EOLAPP1720-P090 EOLAPP2182-P090
Ready to use microbiology media, plated, 90 mm
Media for Microbiology Prepared Media for Microbiology
Manufactured in class 10,000 cleanrooms and quality controlled in compliance with ISO11133-1:20000 and ISO 11133-2:2003 in a UKAS accredited test lab (ISO 17025). Validated according to harmonised pharmacopeia, validation package available on request.
  • Each plate displays: Product code, description, batch number, expiry date, sequential production number, 128 bar code
  • Robust packaging with cut out handles for easy transport; drop tested to ensure maximum protection of product
  • CE marked (if relevant)
  • Pharmaceutical media validated according to harmonised methods
  • Comprehensive product range designed to meet individual customer requirements
Ordering information: Please contact your local Avantor representative if you cannot see the product you are looking for here.