POPSO (β,β'-dihydroxypiperazine-1,4-dipropanesulphonic acid)dihydrate

Supplier: Molekula

16854530-500G 16854530-100G
MOLE16854530-500GEA 451 GBP
MOLE16854530-500G MOLE16854530-100G
POPSO (β,β'-dihydroxypiperazine-1,4-dipropanesulphonic acid)dihydrate
POPSO (β,β'-Dihydroxypiperazine-1,4-dipropanesulphonic acid) dihydrate
Formula: C₁₀H₂₂N₂O₈S₂·2H₂O
Storage Temperature: Ambient
CAS Number: 918131-36-9
EINECS: 269-199-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR