n-Decyl-β-D-maltoside

Supplier: PanReac AppliChem

Synonyms: DM, N-Decyl-Beta-Maltoside, Decyl 4-O-a-D-glucopyranosyl-b-D-glucopyranoside, n-Decyl-beta-D-maltopyranoside, Decyl b-D-maltopyranoside

A6769.0005
A6769.0005EA 1320 GBP
A6769.0005
n-Decyl-β-D-maltoside
n-Decyl-β-D-maltoside
Formula: C₂₂H₄₂O₁₁
MW: 482,56 g/mol
MDL Number: MFCD00061624
CAS Number: 82494-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR