2-Amino-5-bromo-3-(trifluoromethyl)pyridine 96%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 5-Bromo-3-(trifluoromethyl)pyridin-2-amine

H54305.MD H54305.03
H54305.MDEA 0 GBP
H54305.MD H54305.03
2-Amino-5-bromo-3-(trifluoromethyl)pyridine 96%
2-Amino-5-bromo-3-(trifluoromethyl)pyridine

Purity: 96% CAS No.: 79456-34-1 Molecular Formula: C6H4BrF3N2 Formula Weight: 241.01 MDL No.: MFCD10758088

Formula: C₆H₄BrF₃N₂
MW: 241.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10758088
CAS Number: 79456-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR