(±)-Tetrahydrofurfurylamine 97%

Supplier: Thermo Fisher Scientific
Warning

B23068.22 B23068.36
B23068.22EA 23.4 GBP
B23068.22 B23068.36
(±)-Tetrahydrofurfurylamine 97%
(±)-Tetrahydrofurfurylamine

Purity: 97% CAS No.: 4795-29-3 Molecular Formula: C5H11NO Formula Weight: 101.15 Flash Point: 45°(113°F) Refractive Index: 1.4550 Boiling Point: 153-154° Beilstein Reference: 102729 UN No.: 2943 MDL No.: MFCD00005373

Formula: C₅H₁₁NO
Boiling Pt: 153…154 °C
Density: 0,98
Flash Pt: 45 °C (113 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005373
CAS Number: 4795-29-3
EINECS: 225-351-9
UN: 2943
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR