N-Acetylglycinamide(2-acetamidoacetamide) ≥97%

Supplier: Thermo Fisher Scientific

B24940.03 B24940.06 B24940.14
B24940.03EA 49.8 GBP
B24940.03 B24940.06 B24940.14
N-Acetylglycinamide(2-acetamidoacetamide) ≥97%
N-Acetylglycinamide (2-Acetamidoacetamide)

Purity: 97% CAS No.: 2620-63-5 Molecular Formula: CH3CONHCH2CONH2 Formula Weight: 116.12 Odour: Odourless Melting Point: 136-140° Beilstein Reference: 1753982 MDL No.: MFCD00008029

Formula: C₄H₈N₂O₂
Melting Pt: 136…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00008029
CAS Number: 2620-63-5
EINECS: 220-058-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR