Tungsten(VI) ethoxide

Supplier: Thermo Fisher Scientific
Warning

41795.03 41795.06
41795.03EA 68.6 GBP
41795.03 41795.06
Tungsten(VI) ethoxide
Tungsten (VI) ethoxide

CAS No.: 62571-53-3 Molecular Formula: C12H30O6W Formula Weight: 454.21 Physical Form: Powder/chunks Sensitivity: Moisture Sensitive Appearance: Off-white to brown Melting Point: 150° Solubility: Soluble in ethyl acetate UN No.: 3181 MDL No.: MFCD00156526


Intermediate for sol-gel coatings

Formula: C₁₀H₂₅O₅W
Melting Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00156526
CAS Number: 62571-53-3
UN: 3181
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR