2-Amino-6-bromopurine 98%

Supplier: Thermo Fisher Scientific

B21629.06 B21629.03
B21629.06EA 189 GBP
B21629.06 B21629.03
2-Amino-6-bromopurine 98%
2-Amino-6-bromopurine

Purity: 98% CAS No.: 82499-03-4 Molecular Formula: C5H4BrN5 Formula Weight: 214.02 Melting Point: 300° MDL No.: MFCD01321309

Formula: C₅H₄BrN₅
MW: 214.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01321309
CAS Number: 82499-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR