Dimethyldioctadecylammonium bromide (DDA) 99+%

Supplier: Thermo Fisher Scientific
Warning

408260250 408260050 408261000
ACRO408260250EA 436 GBP
ACRO408260250 ACRO408260050 ACRO408261000
Dimethyldioctadecylammonium bromide (DDA) 99+%
Dimethyldioctadecylammonium bromide (DDA)
Formula: C₃₈H₈₀BrN
Melting Pt: 151…157 °C
Density: 1,047 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00041975
CAS Number: 3700-67-2
EINECS: 223-037-6

Order Now

Specification Test Results

Appearance White to almost white Powder and/or chunks
Infrared spectrum Conforms
Melting point ≥150 °C clear point
Titration Argentometric 99.0 to 101.0 %
TLC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR