Ready to use microbiology media, plated, irradiated
Supplier: E AND O LABORATORIES
PP0245-P090
PP8021-P090
EOLAPP0245-P090EA
1.5
GBP
EOLAPP0245-P090
EOLAPP8021-P090
Ready to use microbiology media, plated, irradiated
Media for Microbiology
Prepared Media for Microbiology
Manufactured in class 10,000 cleanrooms and quality controlled in compliance with ISO11133-1:20000 and ISO 11133-2:2003 in a UKAS accredited test lab (ISO 17025). Validated according to harmonised pharmacopeia, validation package available on request.
- Each plate displays: product code, description, batch number, expiry date, sequential production number, 128 bar code
- VHP products resist vapourised hydrogen peroxide
- VHP products can be stored at ambient temperatures
- Quality control tested using recommended strains
- Comprehensive product range designed to meet individual customer requirements
Please contact your local Avantor representative if you can not see the product you are looking for