Ammonium carbamate, Supelco®
Supplier: Merck
|
Danger
|
Synonyms:
Carbamic acid ammonium salt
M-Clarity™ quality level = MQ300
1.01134.0500
1.01134.9050
1.01134.0500EA
157
GBP
1.01134.0500
1.01134.9050
Ammonium carbamate, Supelco®
Ammonium carbamate
Formula:
H₂NCO₂NH₄ MW: 78,07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00013010 CAS Number: 1111-78-0 EINECS: 214-185-2 |
Specification Test Results
Solubility | 790 g/l (20 °C) |
Melting Point | 152 °C (closed capillary tube) |
Molar Mass | 78.07 g/mol |
Bulk Density | 750 kg/m3 |
Vapor Pressure | 82 hPa (20 °C) |
Density | 1.6 g/cm3 (20 °C) |
pH | 10 (100 g/l, H2O, 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...