4-Ethylphenylacetonitrile ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
L10855.03
L10855.06
L10855.03EA
40
GBP
L10855.03
L10855.06
4-Ethylphenylacetonitrile ≥97%
4-Ethylphenylacetonitrile
Purity: 97%
CAS No.: 51632-28-1
Molecular Formula: C10H11N
Formula Weight: 145.21
Refractive Index: 1.5164
Boiling Point: 110°/1mm
UN No.: 3276
MDL No.: MFCD00040893
Formula:
C₁₀H₁₁N Boiling Pt: 110 °C (1 mmHg) Density: 0,970 Storage Temperature: Ambient |
MDL Number:
MFCD00040893 CAS Number: 51632-28-1 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...