Cacodylic acid sodium salt trihydrate

Supplier: Molekula
Danger

Synonyms: Dimethylarsonic acid sodium salt trihydrateSodium cacodylate trihydrateSodium dimethylarsinate trihydrateDimethylarsinic acid sodium salt trihydrate

MOLEM55777472EA 0 GBP
MOLEM55777472
Cacodylic acid sodium salt trihydrate
Cacodylic acid sodium salt trihydrate
Formula: (CH₃)₂As(O)ONa·3H₂O
MW: 213,94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149079
CAS Number: 6131-99-3
EINECS: 204-708-2
UN: 1688
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR