2-Chloro-5-pentylpyrimidine ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
H27808.09
H27808.03
H27808.09EA
79.3
GBP
H27808.09
H27808.03
2-Chloro-5-pentylpyrimidine ≥98%
2-Chloro-5-pentylpyrimidine
Purity: 98%
CAS No.: 154466-62-3
Molecular Formula: C9H13ClN2
Formula Weight: 184.67
Flash Point: 97°(207°F)
Refractive Index: 1.504
Boiling Point: 124°/3mm
MDL No.: MFCD04038093
Formula:
C₉H₁₃ClN₂ MW: 184.67 g/mol Boiling Pt: 124 °C (3 mmHg) Density: 1.085 Flash Pt: 97 °C (207 °F) Storage Temperature: Ambient |
MDL Number:
MFCD04038093 CAS Number: 154466-62-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...