Quinuclidine ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
1-Azabicyclo[2.2.2]octane, ABCO
H54498.MD
H54498.03
H54498.06
H54498.MDEA
66
GBP
H54498.MD
H54498.03
H54498.06
Quinuclidine ≥97%
Quinuclidine
Formula:
C₇H₁₃N MW: 111,19 g/mol Boiling Pt: 150 °C (1013 hPa) Melting Pt: 157…160 °C Density: 1,025 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006690 CAS Number: 100-76-5 EINECS: 202-887-1 UN: 2811 ADR: 6.1,II Merck Index: 14,8081 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...