1,2,3-Thiadiazole-4-carboxylic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
B20842.03
B20842.06
B20842.03EA
140
GBP
B20842.03
B20842.06
1,2,3-Thiadiazole-4-carboxylic acid ≥97%
1,2,3-Thiadiazole-4-carboxylic acid
Purity: 97%
CAS No.: 4100-13-4
Molecular Formula: C3H2N2O2S
Formula Weight: 130.12
Melting Point: 227-228°
Beilstein Reference: 113707
Formula:
C₃H₂N₂O₂S Melting Pt: 227…228 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052099 CAS Number: 4100-13-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...