3,5-Diphenyl-1H-pyrazole 99%

Supplier: Thermo Scientific

337870050 337870250
ACRO337870050EA 98.4 GBP
ACRO337870050 ACRO337870250
3,5-Diphenyl-1H-pyrazole 99%
3,5-Diphenyl-1H-pyrazole
Formula: C₁₅H₁₂N₂
MW: 220,27 g/mol
Melting Pt: 200…203 °C
Storage Temperature: Ambient
MDL Number: MFCD00039675
CAS Number: 1145-01-3
EINECS: 214-543-8

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 200 to 204 °C
HPLC ≥98.5 %
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR