Quinazoline ≥98%

Supplier: Thermo Scientific

B24094.03 B24094.06
B24094.03EA 26 GBP
B24094.03 B24094.06
Quinazoline ≥98%
Quinazoline

Purity: 98% CAS No.: 253-82-7 Molecular Formula: C8H6N2 Formula Weight: 130.15 Flash Point: 106°(224°F) Melting Point: 45-48° Boiling Point: 243° Merck Reference: 14,8053 MDL No.: MFCD00006712

Formula: C₈H₆N₂
MW: 130,15 g/mol
Boiling Pt: 243 °C
Melting Pt: 45…48 °C
Flash Pt: 106 °C (224 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006712
CAS Number: 253-82-7
EINECS: 205-965-3
Merck Index: 14,8053

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR