3-Amino-1H-pyrazole ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 3-Pyrazolamine, 3-AP, 1H-Pyrazol-3-amine, 3-Aminopyrazole

A14458.03 A14458.06 A14458.14
A14458.03EA 37.6 GBP
A14458.03 A14458.06 A14458.14
3-Amino-1H-pyrazole ≥97%
3-Amino-1H-pyrazole

Purity: 97+% CAS No.: 1820-80-0 Molecular Formula: C3H5N3 Formula Weight: 83.09 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 34-39° Boiling Point: 218°/122mm Beilstein Reference: 1618 UN No.: 3259 MDL No.: MFCD00005236

Formula: C₃H₅N₃
MW: 83,09 g/mol
Boiling Pt: 218 °C (122 mmHg)
Melting Pt: 34…39 °C
Flash Pt: >110 °C (230 °F)
MDL Number: MFCD00005236
CAS Number: 1820-80-0
EINECS: 217-338-1
UN: 3259
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR