Denatonium benzoate
Supplier: Molekula
Synonyms:
N-Benzyl-2-((2,6-dimethylphenyl)amino)-N,N-diethyl-2-oxoethanaminium benzoate
MOLE90005192-5GEA
0
GBP
MOLE90005192-5G
Denatonium benzoate
Denatonium benzoate
Formula:
C₂₈H₃₄N₂O₃ MW: 446,59 g/mol |
MDL Number:
MFCD00031578 CAS Number: 3734-33-6 EINECS: 223-095-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...