1-Benzyl-2-pyrrolidinone 97%

Supplier: Thermo Scientific

Synonyms: 1-Benzylpyrrolidin-2-one

154120250 154121000
ACRO154120250EA 81.3 GBP
ACRO154120250 ACRO154121000
1-Benzyl-2-pyrrolidinone 97%
1-Benzyl-2-pyrrolidinone
Formula: C₁₁H₁₃NO
MW: 175,23 g/mol
Density: 1,09 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00003195
CAS Number: 5291-77-0
EINECS: 226-131-5

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5500 to 1.5530 (20°C, 589 nm)
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR