4-Tritylaniline ≥97%

Supplier: Thermo Scientific

Synonyms: 4-(Triphenylmethyl)aniline

A10550.06 A10550.14
A10550.06EA 32.5 GBP
A10550.06 A10550.14
4-Tritylaniline ≥97%
4-Tritylaniline

Purity: 97% CAS No.: 22948-06-7 Molecular Formula: C25H21N Formula Weight: 335.45 Melting Point: 256-260° Beilstein Reference: 2750328

Formula: C₂₅H₂₁N
MW: 335,45 g/mol
Melting Pt: 256…260 °C
Storage Temperature: Ambient
MDL Number: MFCD00007898
CAS Number: 22948-06-7
EINECS: 245-346-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR