2-Benzylpyridine ≥98%
Supplier: Thermo Scientific
|
Warning
|
A10662.18
A10662.0B
A10662.30
A10662.18EA
24.6
GBP
A10662.18
A10662.0B
A10662.30
2-Benzylpyridine ≥98%
2-Benzylpyridine
Purity: 98+%
CAS No.: 101-82-6
Molecular Formula: C12H11N
Formula Weight: 169.23
Flash Point: 125°(257°F)
Refractive Index: 1.5790
Melting Point: 10-12°
Boiling Point: 140-143°/11mm
Beilstein Reference: 115875
MDL No.: MFCD00006352
Formula:
C₁₂H₁₁N MW: 169,23 g/mol Boiling Pt: 140…143 °C (11 mmHg) Melting Pt: 10…12 °C Density: 1,054 Flash Pt: 125 °C (257 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006352 CAS Number: 101-82-6 EINECS: 202-979-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...