Pentachloropyridine
Supplier: Molekula
MOLEM22033184EA
0
GBP
MOLEM22033184
Pentachloropyridine
Pentachloropyridine
Formula:
C₅Cl₅N MW: 251,33 g/mol Boiling Pt: 279…280 °C (1013 hPa) Melting Pt: 124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006230 CAS Number: 2176-62-7 EINECS: 218-535-5 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...