1-Bromo-4-fluoronaphthalene ≥98%
Supplier: Thermo Scientific
|
Warning
|
A16934.06
A16934.03
A16934.14
A16934.06EA
116
GBP
A16934.06
A16934.03
A16934.14
1-Bromo-4-fluoronaphthalene ≥98%
1-Bromo-4-fluoronaphthalene
Purity: 98%
CAS No.: 341-41-3
Molecular Formula: C10H6BrF
Formula Weight: 225.06
Flash Point:
Formula:
C₁₀H₆BrF MW: 225,06 g/mol Melting Pt: 33…37 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051473 CAS Number: 341-41-3 EINECS: 206-434-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...