Zirconium(IV) oxide, Patinal®
Supplier: Merck
|
Warning
|
Synonyms:
Zirconium dioxide, Zirconium Oxide
1.08905.1000
1.08905.0100
1.08905.1000EA
0
GBP
1.08905.1000
1.08905.0100
Zirconium(IV) oxide, Patinal®
Zirconium (IV) oxide
Formula:
ZrO₂ MW: 123,22 g/mol Boiling Pt: 4300 °C (1013 hPa) Melting Pt: 2680 °C Density: 5,75 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011310 CAS Number: 1314-23-4 EINECS: 215-227-2 Merck Index: 14,10180 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...