Lanthanum(III) fluoride, Patinal®

Supplier: Merck

Synonyms: Lanthanum trifluoride

1.01129.0100 1.01129.1000
1.01129.0100EA 0 GBP
1.01129.0100 1.01129.1000
Lanthanum(III) fluoride, Patinal®
Lanthanum (III) fluoride
Formula: F₃La
MW: 195,9 g/mol
Boiling Pt: ∼2327 °C (1013 hPa)
Melting Pt: 1491,85…1507.85 °C
Density: 5,936 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011073
CAS Number: 13709-38-1
EINECS: 237-252-8
UN: 3288
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR