Sodium-3-nitrobenzenesulphonate

Supplier: Molekula

Synonyms: Sodium 3-nitrobenzenesulphonate3-Nitrobenzenesulphonic acid, sodium salt

MOLEM84233913EA 0 GBP
MOLEM84233913
Sodium-3-nitrobenzenesulphonate
Sodium 3-nitrobenzenesulphonate
Formula: C₆H₄NNaO₅S
MW: 225,16 g/mol
MDL Number: MFCD00007490
CAS Number: 127-68-4
EINECS: 204-857-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR