3-Methyl-4-phenyl-1H-pyrazole ≥98%
Supplier: Thermo Scientific
|
Warning
|
L07250.03
L07250.06
L07250.03EA
83.3
GBP
L07250.03
L07250.06
3-Methyl-4-phenyl-1H-pyrazole ≥98%
3-Methyl-4-phenyl-1H-pyrazole
Purity: 98%
CAS No.: 13788-84-6
Molecular Formula: C10H10N2
Formula Weight: 158.20
Melting Point: 140-144°
Beilstein Reference: 3330
Formula:
C₁₀H₁₀N₂ Melting Pt: 140…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051752 CAS Number: 13788-84-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...