6-Aminopicolinic acid 95%

Supplier: Thermo Scientific
Warning

Synonyms: 6-Aminopyridine-2-carboxylic acid

H31861.03 H31861.09
H31861.03EA 44.7 GBP
H31861.03 H31861.09
6-Aminopicolinic acid 95%
6-Aminopicolinic acid

Purity: 95% CAS No.: 23628-31-1 Molecular Formula: C6H6N2O2 Formula Weight: 138.13 Odour: Odourless Melting Point: ca 295° dec. MDL No.: MFCD00233711

Formula: C₆H₆N₂O₂
MW: 138,13 g/mol
Melting Pt: ∼ 295 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00233711
CAS Number: 23628-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR