3-Sulpholene 98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
3-sulpholene, Butadiene sulphone
107611000
107610025
107615000
ACRO107611000EA
38.4
GBP
ACRO107611000
ACRO107610025
ACRO107615000
3-Sulpholene 98%
3-Sulpholene
Formula:
C₄H₆O₂S MW: 118,16 g/mol Melting Pt: 63…66 °C Flash Pt: 112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005481 CAS Number: 77-79-2 EINECS: 201-059-7 UN: 3335 ADR: 9,III |
Specification Test Results
Appearance | White to off-white Adhering powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 63 to 67 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...