Phosphorous acid 98
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Phosphonic acid
A11189.0B
A11189.0I
A11189.30
A11189.0BEA
93.6
GBP
A11189.0B
A11189.0I
A11189.30
Phosphorous acid 98
Phosphorous acid
Purity: 98+%
CAS No.: 13598-36-2
Molecular Formula: H3PO3
Formula Weight: 82.00
Odour: Odourless
Melting Point: 74°
Boiling Point: 200° dec.
Merck Reference: 14,7346
UN No.: 2834
MDL No.: MFCD00137258
Formula:
H₃O₃P MW: 82 g/mol Boiling Pt: 200 °C (dec.) Density: 1.651 Storage Temperature: Ambient |
MDL Number:
MFCD00137258 CAS Number: 13598-36-2 EINECS: 237-066-7 UN: 2834 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...