Sodium cyanate 95%

Supplier: Thermo Scientific
Warning

A12413.0E A12413.22 A12413.36
A12413.0EEA 126 GBP
A12413.0E A12413.22 A12413.36
Sodium cyanate 95%
Sodium cyanate

Purity: 95% CAS No.: 917-61-3 Molecular Formula: NaOCN Formula Weight: 65.01 Melting Point: 550° Beilstein Reference: 3594963 Merck Reference: 14,8604 MDL No.: MFCD00011121

Formula: CNNaO
MW: 65,01 g/mol
Melting Pt: 550 °C
Density: 1,89
Storage Temperature: Ambient
MDL Number: MFCD00011121
CAS Number: 917-61-3
EINECS: 213-030-6
Merck Index: 14,8604

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR