Potassium iodate, Supelco®
Supplier: Merck
|
Warning
|
SUPL60386-50G-FEA
276
GBP
SUPL60386-50G-F
NOVS60386-50G-F
Potassium iodate, Supelco®
Potassium iodate
Formula:
KlO₃ MW: 214 g/mol Melting Pt: 560 °C Density: 3,98 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011406 CAS Number: 7758-05-6 EINECS: 231-831-9 UN: 1479 ADR: 5.1,II Merck Index: 12,07808 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...