Magnesium ethanolate, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Magnesium ethoxide, Magnesium ethylate

M-Clarity™ quality level = MQ200

8.18894.0100 8.18894.0500
8.18894.0100EA 90.4 GBP
8.18894.0100 8.18894.0500
Magnesium ethanolate, Sigma-Aldrich®
Magnesium ethanolate
Formula: C₄H₁₀MgO₂
MW: 114,43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009060
CAS Number: 2414-98-4
EINECS: 219-323-5
UN: 3205
ADR: 4.2,II

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Molar Mass 114.43 g/mol
Bulk Density 480 kg/m3
Ignition Point 70 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR