Benzoyl peroxide stabilised for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Dibenzoyl peroxide

Stabilisation: Moistened with water 25%
M-Clarity™ quality level = MQ200

8.01641.0100 8.01641.0250 8.01641.1000 8.01641.9026 8.01641.0010
8.01641.0100EA 23.8 GBP
8.01641.0100 8.01641.0250 8.01641.1000 8.01641.9026 8.01641.0010
Benzoyl peroxide stabilised for synthesis, Sigma-Aldrich®
Benzoyl peroxide
Formula: C₁₄H₁₀O₄
MW: 242,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003071
CAS Number: 94-36-0
EINECS: 202-327-6
UN: 3104
ADR: 5.2,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR