2-Amino-3-formylpyridine 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-Amino-3-pyridinecarboxaldehyde, 2-Aminonicotinaldehyde, 2-Aminopyridine-3-carboxaldehyde
L19620.03
L19620.06
L19620.03EA
56.4
GBP
L19620.03
L19620.06
2-Amino-3-formylpyridine 98%
2-Amino-3-formylpyridine
Purity: 98%
CAS No.: 7521-41-7
Molecular Formula: C6H6N2O
Formula Weight: 122.13
Storage Temperature: 2°C to 8°C
Melting Point: 99-102°
Beilstein Reference: 109598
MDL No.: MFCD01830382
Formula:
C₆H₆N₂O Melting Pt: 99…102 °C |
MDL Number:
MFCD01830382 CAS Number: 7521-41-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...