D-(-)-Quinic acid ≥98%

Supplier: Thermo Scientific

Synonyms: (1α,3R,4α,5R)-1,3,4,5-Tetrahydroxycyclohexanecarboxylic acid

L15238.06 L15238.14
L15238.06EA 19.5 GBP
L15238.06 L15238.14
D-(-)-Quinic acid ≥98%
D(-)-Quinic acid

Purity: 98% CAS No.: 77-95-2 Molecular Formula: C7H12O6 Formula Weight: 192.17 Odour: Odourless Melting Point: 162-167° Beilstein Reference: 2212412 Merck Reference: 14,8059 Optical Rotation: -44° (c=20 in water) MDL No.: MFCD00003864

Formula: C₇H₁₂O₆
MW: 192,17 g/mol
Melting Pt: 162…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00003864
CAS Number: 77-95-2
EINECS: 201-072-8
Merck Index: 14,8059

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR