Piperidine-N-carbaldehyde ≥99%

Supplier: Thermo Scientific
Danger

Synonyms: 1-Formylpiperidine, N-Formylpiperidine

B23452.30 B23452.0B B23452.18
B23452.30EA 103 GBP
B23452.30 B23452.0B B23452.18
Piperidine-N-carbaldehyde ≥99%
Piperidine-N-carbaldehyde

Purity: 99% CAS No.: 2591-86-8 Molecular Formula: C6H11NO Formula Weight: 113.16 Flash Point: 102°(215°F) Refractive Index: 1.4840 Melting Point: -31° Boiling Point: 222° Beilstein Reference: 107697 MDL No.: MFCD00006483

Formula: C₆H₁₁NO
MW: 113,16 g/mol
Boiling Pt: 222 °C
Melting Pt: –31 °C
Density: 1,019
Flash Pt: 102 °C (215 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006483
CAS Number: 2591-86-8
EINECS: 219-986-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR