Vinyl benzoate 95%
Supplier: Thermo Fisher Scientific
|
Warning
|
B21805.22
B21805.36
B21805.22EA
65.2
GBP
B21805.22
B21805.36
Vinyl benzoate 95%
Vinyl benzoate
Purity: 95%
CAS No.: 769-78-8
Molecular Formula: C9H8O2
Formula Weight: 148.16
Flash Point: 82°(180°F)
Refractive Index: 1.529
Boiling Point: 73-74°/8mm
Beilstein Reference: 2041125
MDL No.: MFCD00048141
Formula:
C₉H₈O₂ MW: 148.16 g/mol Boiling Pt: 73…74 °C (8 mmHg) Density: 1.070 Flash Pt: 82 °C (180 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00048141 CAS Number: 769-78-8 EINECS: 212-214-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...