2-Furoyl chloride 98+%
Supplier: Thermo Scientific
|
Danger
|
165381000
165380250
ACRO165381000EA
128
GBP
ACRO165381000
ACRO165380250
2-Furoyl chloride 98+%
2-Furoyl chloride
Formula:
C₅H₃ClO₂ MW: 130,53 g/mol Boiling Pt: 173…174 °C (760 mmHg) Melting Pt: –2 °C Density: 1,32 g/cm³ Flash Pt: 85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003228 CAS Number: 527-69-5 EINECS: 208-422-9 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to yellow to light orange Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5300 to 1.5320 (20°C, 589 nm) |
GC | ≥98.0 % |
Titration RCOCl | ≥98.0 % |
Impurity | ≤2.0 % Free carboxylic acid (as furoic acid) |
Free acid (HCl) | ≤0.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...