D-(-)-Quinic acid 98+%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(1α,3R,4α,5R)-1,3,4,5-Tetrahydroxycyclohexanecarboxylic acid
160500250
160501000
160505000
ACRO160500250EA
72.5
GBP
ACRO160500250
ACRO160501000
ACRO160505000
D-(-)-Quinic acid 98+%
D(-)-Quinic acid
Formula:
C₇H₁₂O₆ MW: 192,17 g/mol Melting Pt: 166…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003864 CAS Number: 77-95-2 EINECS: 201-072-8 |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 162 to 168 °C |
Specific optical rotation | -42° to -45° (20 °C, 589 nm) (c=10, water) |
Titration with NaOH | ≥98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...