4-Chloro-6-fluoro-2-methylquinoline 96%

Supplier: Thermo Scientific
Warning

H50553.03 H50553.MD
H50553.03EA 280 GBP
H50553.03 H50553.MD
4-Chloro-6-fluoro-2-methylquinoline 96%
4-Chloro-6-fluoro-2-methylquinoline

Purity: 96% CAS No.: 18529-01-6 Molecular Formula: C10H7ClFN Formula Weight: 195.63 MDL No.: MFCD00272332

Formula: C₁₀H₇ClFN
MW: 195,62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00272332
CAS Number: 18529-01-6
EINECS: 242-403-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR